Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012010.3 | NP_036140.1 | 12 | 2788 | ||
GenBank mRNA | AK128949.1 | - | 12 | 2786 | ||
AK145741.1 | - | 3 | 1715 | |||
AK146900.1 | - | 12 | 2829 | |||
AK150095.1 | - | 12 | 2796 | |||
AK151579.1 | - | 8 | 2344 | |||
AK153568.1 | - | 8 | 2344 | |||
AK165866.1 | - | 12 | 2790 | |||
BC006682.1 | - | 1 | 418 | |||
BC036993.1 | - | 12 | 2786 | |||
BC063755.1 | - | 12 | 2788 | 6513251 | ||
BC066793.1 | - | 2 | 1526 |