Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012024.2 | NP_036154.1 | 12 | 1750 | ||
GenBank mRNA | AK053228.1 | - | 11 | 1601 | ||
AK088366.1 | - | 12 | 1722 | |||
AK132432.1 | - | 12 | 1783 | |||
AK133824.1 | - | 11 | 1693 | |||
AK162184.1 | - | 3 | 261 | |||
AK220163.1 | - | 12 | 1740 | |||
BC019199.1 | - | 6 | 475 | |||
BC085149.1 | - | 12 | 1750 | 6848033 | ||
BC094387.1 | - | 12 | 1740 | |||
U49728.1 | - | 10 | 1030 |