Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253808.1 | NP_001240737.1 | 3 | 378 | ||
NM_001253809.1 | NP_001240738.1 | 3 | 374 | |||
NM_012025.7 | NP_036155.1 | 4 | 509 | |||
GenBank mRNA | AB030252.1 | - | 3 | 358 | ||
AF079974.1 | - | 3 | 319 | |||
AF212320.1 | - | 3 | 345 | |||
AF212321.1 | - | 4 | 476 | |||
AK053850.1 | - | 3 | 339 | |||
AK144608.1 | - | 4 | 477 | |||
AK144745.1 | - | 3 | 337 | |||
AK154929.1 | - | 4 | 497 | |||
AK162660.1 | - | 3 | 305 | |||
AK165897.1 | - | 3 | 331 | |||
AK166084.1 | - | 4 | 467 | |||
AK166094.1 | - | 3 | 336 | |||
AK168020.1 | - | 3 | 335 | |||
AK168025.1 | - | 3 | 335 | |||
AK168170.1 | - | 3 | 335 | |||
AK168679.1 | - | 3 | 319 | |||
BC010715.1 | - | 3 | 324 | 3602242 |