Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012028.4 | NP_036158.3 | 5 | 1560 | ||
GenBank mRNA | AB028840.1 | - | 1543 | |||
AB030836.1 | - | 5 | 1563 | |||
AK034387.1 | - | 1564 | ||||
AK038434.1 | - | 5 | 1564 | |||
AK042683.1 | - | 5 | 1600 | |||
BC055737.1 | - | 5 | 1560 | 6415191 |