Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012033.3 | NP_036163.3 | 3 | 521 | ||
GenBank mRNA | AF153366.1 | - | 3 | 463 | ||
AK008374.1 | - | 3 | 489 | |||
BC010745.1 | - | 3 | 466 | 4239192 |