Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012034.3 | NP_036164.2 | 3 | 408 | ||
GenBank mRNA | AF175523.1 | - | 3 | 309 | ||
AK027982.1 | - | 2 | 234 | |||
BC019974.1 | - | 3 | 332 | 4215589 | ||
BC052325.1 | - | 3 | 377 | 6531021 |