Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012034.3 | NP_036164.2 | 4 | 661 | ||
GenBank mRNA | AF175523.1 | - | 4 | 562 | ||
AK027982.1 | - | 3 | 487 | |||
BC019974.1 | - | 4 | 585 | 4215589 | ||
BC052325.1 | - | 4 | 630 | 6531021 |