Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zw10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012039.2 | NP_036169.1 | 1, 2 | 215 | ||
GenBank mRNA | AK145270.1 | - | 1, 2 | 184 | ||
AK167251.1 | - | 1, 2 | 184 | |||
AK167937.1 | - | 1, 2 | 144 | |||
AK168479.1 | - | 1, 2 | 184 | |||
BC012435.1 | - | 1, 2 | 164 | 3711269 | ||
CJ172982.1 | - | 1, 2 | 216 |