Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zw10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012039.2 | NP_036169.1 | 5 | 536 | ||
GenBank mRNA | AK018243.1 | - | 4 | 335 | ||
AK167251.1 | - | 5 | 505 | |||
AK167937.1 | - | 5 | 465 | |||
AK168479.1 | - | 5 | 505 | |||
BC012435.1 | - | 5 | 485 | 3711269 | ||
BC067405.1 | - | 2575 |