Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242580.1 | NP_001229509.1 | 7 | 1199 | ||
NM_012049.2 | NP_036179.1 | 7 | 1167 | |||
GenBank mRNA | AF069988.1 | - | 7 | 1148 | ||
AK002269.1 | - | 7 | 1207 | |||
AK004988.2 | - | 1160 | ||||
AK032189.1 | - | 1850 | ||||
AK203644.1 | - | 1 | 254 |