Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012052.2 | NP_036182.1 | 5 | 582 | ||
GenBank mRNA | AK003181.1 | - | 5 | 520 | ||
AK010579.1 | - | 5 | 525 | |||
AK010678.1 | - | 5 | 523 | |||
AK011655.1 | - | 5 | 520 | |||
AK012234.1 | - | 5 | 524 | |||
AK012314.1 | - | 5 | 524 | |||
AK012538.1 | - | 5 | 525 | |||
AK051243.1 | - | 5 | 525 | |||
AK088038.1 | - | 5 | 526 | |||
AK143603.1 | - | 4 | 2157 | |||
AK146796.1 | - | 5 | 525 | |||
AK148030.1 | - | 5 | 524 | |||
AK150479.1 | - | 5 | 518 | |||
AK159508.1 | - | 5 | 525 | |||
AK160526.1 | - | 5 | 524 | |||
AK164077.1 | - | 5 | 525 | |||
AK165174.1 | - | 5 | 514 | |||
AK166750.1 | - | 5 | 526 | |||
AK167210.1 | - | 5 | 524 | |||
AK167266.1 | - | 5 | 514 | |||
AK168845.1 | - | 5 | 514 | |||
BC010721.1 | - | 5 | 512 | 2811930 | ||
U89414.1 | - | 3 | 300 | |||
X76772.1 | - | 5 | 499 |