Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012056.2 | NP_036186.2 | 3, 4 | 713 | ||
GenBank mRNA | AF090334.1 | - | 3, 4 | 639 | ||
AF279263.1 | - | 3, 4 | 691 | |||
AK007499.1 | - | 1, 2 | 103 | |||
BC026133.1 | - | 3, 4 | 653 | 4018235 | ||
BC043129.1 | - | 3, 4 | 713 | 6810029 |