Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082414.2 | NP_001075883.2 | 11 | 1973 | ||
GenBank mRNA | AF003234.1 | - | 5 | 363 | ||
AK050185.1 | - | 3 | 230 | |||
BC028254.1 | - | 11 | 1521 | |||
BC031117.1 | - | 11 | 1084 | 5064197 | ||
D89677.2 | - | 17 | 2459 |