Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161730.1 | NP_001155202.1 | 4 | 1142 | ||
NM_013683.2 | NP_038711.2 | 4 | 1142 | |||
GenBank mRNA | AK040629.1 | - | 4 | 971 | ||
AK040919.1 | - | 2 | 2105 | |||
AK151481.1 | - | 4 | 897 | |||
AK152953.1 | - | 4 | 842 | |||
AK153890.1 | - | 4 | 1146 | |||
AK162731.1 | - | 4 | 942 | |||
AK166046.1 | - | 4 | 1143 | |||
AK167485.1 | - | 4 | 1144 | |||
AK171363.1 | - | 4 | 897 | |||
BC013802.1 | - | 4 | 553 | 3257566 | ||
BC024897.1 | - | 4 | 665 | 3257734 | ||
BC031584.1 | - | 1 | 717 | |||
M55637.1 | - | 4 | 534 | |||
U60018.1 | - | 4 | 818 | |||
U60019.1 | - | 4 | 818 | |||
U60020.1 | - | 4 | 818 | |||
U60021.1 | - | 4 | 818 | |||
U60022.1 | - | 4 | 818 | |||
U60023.1 | - | 4 | 818 | |||
X59615.1 | - | 4 | 377 |