Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083967.1 | NP_001077436.1 | 11 | 1147 | ||
NM_013685.2 | NP_038713.1 | 11 | 1462 | |||
GenBank mRNA | AK014343.1 | - | 4 | 531 | ||
AK028748.1 | - | 11 | 1073 | |||
AK081012.1 | - | 11 | 1099 | |||
AK132140.1 | - | 4 | 488 | |||
AK133885.1 | - | 12 | 1874 | |||
AK151433.1 | - | 4 | 433 | |||
BC014293.1 | - | 4 | 484 | 4014231 | ||
BC043050.1 | - | 10 | 922 | 5708370 | ||
S75870.1 | - | 11 | 1034 | |||
U16321.1 | - | 5 | 521 | |||
U16322.1 | - | 12 | 1434 | |||
X91753.1 | - | 11 | 1086 |