Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013687.3 | NP_038715.2 | 2 | 213 | ||
GenBank mRNA | AK053725.1 | - | 1 | 425 | ||
BC016610.1 | - | 2 | 196 | |||
BC058793.1 | - | 1 | 152 | |||
BC089527.1 | - | 2 | 214 | |||
X52128.1 | - | 2 | 193 |