Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uncx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013702.3 | NP_038730.1 | 3 | 1772 | ||
GenBank mRNA | AF247550.1 | - | 3 | 1761 | ||
AJ001116.1 | - | 3 | 1732 | |||
BC051973.1 | - | 3 | 1734 | 5718115 |