Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013706.2 | NP_038734.1 | 2 | 429 | ||
GenBank mRNA | AK088161.1 | - | 2 | 396 | ||
AK155728.1 | - | 2 | 430 | |||
BC027495.1 | - | 2 | 379 | 1396480 | ||
M55561.1 | - | 2 | 418 | |||
X56745.1 | - | 1 | 173 | |||
X76696.1 | - | 2 | 405 |