Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177701.2 | NP_001171172.1 | 4 | 638 | ||
NM_006860.4 | NP_006851.1 | 4 | 635 | |||
XM_006724106.2 | XP_006724169.1 | 5 | 399 | |||
XM_017028540.1 | XP_016884029.1 | 4 | 642 | |||
GenBank mRNA | AB209424.1 | - | 4 | 630 | ||
AL022729.1 | - | 4 | 579 | |||
BC000566.2 | - | 4 | 616 | 3163341 | ||
BM711121.1 | - | 4 | 226 | |||
BT006815.1 | - | 4 | 212 | |||
CR456558.1 | - | 4 | 254 |