Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177806.1 | NP_001171277.1 | 22 | 3071 | ||
NM_013719.3 | NP_038747.2 | 23 | 3375 | |||
GenBank mRNA | AF193342.1 | - | 18 | 3278 | ||
AF193343.1 | - | 23 | 3367 | |||
AF193344.1 | - | 23 | 3459 | |||
AJ243533.1 | - | 23 | 3375 | |||
AK077199.1 | - | 8 | 887 | |||
AK129334.1 | - | 18 | 2725 | |||
BC072637.1 | - | 22 | 3071 | 30354433 | ||
CF748039.1 | - | 6 | 541 |