Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190717.1 | NP_001177646.1 | 3 | 489 | ||
NM_013726.3 | NP_038754.1 | 3 | 489 | |||
GenBank mRNA | AJ003132.1 | - | 3 | 379 | ||
AK014480.1 | - | 3 | 455 | |||
AK132403.1 | - | 3 | 454 | |||
AK163408.1 | - | 1 | 65 | |||
BB611145.1 | - | 3 | 490 | |||
BC069983.1 | - | 3 | 437 | |||
BC138709.1 | - | 3 | 307 | 8861731 | ||
BC138710.1 | - | 3 | 307 | 8861732 | ||
BC145392.1 | - | 3 | 307 | 9053950 |