Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290830.1 | NP_001277759.1 | 7 | 869 | ||
NM_013735.4 | NP_038763.3 | 7 | 869 | |||
GenBank mRNA | AJ414734.1 | - | 6 | 730 | ||
AK045851.1 | - | 7 | 784 | |||
AK148123.1 | - | 7 | 829 | |||
BC079906.1 | - | 7 | 803 | 30639375 |