Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrasls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013751.5 | NP_038779.2 | 4 | 1889 | ||
GenBank mRNA | AK012719.1 | - | 1 | 897 | ||
AK039471.1 | - | 4 | 1891 |