Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289898.1 | NP_001276827.1 | 11 | 2440 | ||
NM_013767.6 | NP_038795.3 | 11 | 2444 | |||
GenBank mRNA | AK035978.1 | - | 5 | 1540 | ||
BC026127.1 | - | 11 | 2384 | 4010696 |