Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013771.5 | NP_038799.1 | 19 | 2625 | ||
GenBank mRNA | AF090430.1 | - | 19 | 2603 | ||
AK021130.1 | - | 1 | 42 | |||
AK045141.1 | - | 19 | 2609 | |||
AK079076.1 | - | 2 | 450 | |||
AK079175.1 | - | 19 | 2585 | |||
BC007128.1 | - | 19 | 2586 | 3591123 |