Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013786.2 | NP_038814.1 | 4 | 612 | ||
GenBank mRNA | AK008212.1 | - | 4 | 671 | ||
BC021836.1 | - | 4 | 612 | 5135142 |