Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013788.2 | NP_038816.1 | 1 | 2551 | ||
GenBank mRNA | AF148857.1 | - | 1 | 2550 | ||
AK142886.1 | - | 1 | 1334 |