Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044747.2 | NP_001038212.1 | 5 | 798 | ||
NM_001163797.1 | NP_001157269.1 | 6 | 888 | |||
NM_013844.2 | NP_038872.1 | 6 | 894 | |||
GenBank mRNA | AB024005.1 | - | 6 | 878 | ||
AK013868.1 | - | 6 | 889 | |||
AK044831.1 | - | 5 | 768 | |||
AK146092.1 | - | 6 | 855 | |||
AK153887.1 | - | 6 | 830 | |||
BC018171.1 | - | 6 | 877 | 3989444 |