Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ror1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013845.4 | NP_038873.2 | 3 | 612 | ||
GenBank mRNA | AB010383.1 | - | 3 | 184 | ||
AK046699.1 | - | 3 | 474 | |||
AK049369.1 | - | 3 | 613 | |||
AK080896.1 | - | 3 | 560 | |||
AK087548.1 | - | 3 | 543 | |||
BC095933.1 | - | 3 | 561 | |||
BC138794.1 | - | 3 | 597 | 8861816 | ||
BC138810.1 | - | 3 | 597 | 8861832 |