Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ror1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013845.4 | NP_038873.2 | 5 | 959 | ||
GenBank mRNA | AB010383.1 | - | 5 | 531 | ||
AK046699.1 | - | 5 | 821 | |||
AK049369.1 | - | 5 | 960 | |||
AK080896.1 | - | 5 | 907 | |||
AK087548.1 | - | 5 | 890 | |||
BC138794.1 | - | 5 | 944 | 8861816 | ||
BC138810.1 | - | 5 | 944 | 8861832 |