Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006891.3 | NP_008822.2 | 3 | 422 | ||
GenBank mRNA | BC117338.1 | - | 3 | 422 | 40125889 | |
BC117340.1 | - | 3 | 422 | 40125891 | ||
CD674533.1 | - | 2 | 286 | |||
EL946552.1 | - | 3 | 338 | |||
KT966392.1 | - | 3 | 330 | |||
U66583.1 | - | 2 | 306 |