Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013846.3 | NP_038874.3 | 7 | 1389 | ||
GenBank mRNA | AB010384.1 | - | 7 | 1157 | ||
AK031112.1 | - | 7 | 1359 | |||
AK031789.1 | - | 7 | 1147 | |||
AK083538.1 | - | 7 | 1365 | |||
AK084752.1 | - | 7 | 1355 | |||
AK157705.1 | - | 7 | 1405 | |||
BC030848.1 | - | 5 | 730 | |||
BC070436.1 | - | 7 | 1389 | 30535615 |