Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161712.1 | NP_001155184.1 | 4 | 569 | ||
NM_013847.4 | NP_038875.3 | 4 | 569 | |||
GenBank mRNA | AF093403.1 | - | 4 | 527 | ||
AK007395.1 | - | 4 | 492 | |||
AK013138.1 | - | 4 | 524 | |||
AK075782.1 | - | 4 | 509 | |||
AK076469.1 | - | 4 | 542 | |||
AK087433.1 | - | 4 | 528 | |||
BC024107.1 | - | 4 | 569 | 5125361 |