Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013847.4 | NP_038875.3 | 9 | 1239 | ||
GenBank mRNA | AF093403.1 | - | 9 | 1197 | ||
AK007395.1 | - | 9 | 1172 | |||
AK013138.1 | - | 9 | 1194 | |||
AK075782.1 | - | 9 | 1179 | |||
AK087433.1 | - | 9 | 1198 | |||
BC024107.1 | - | 9 | 1239 | 5125361 |