Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013851.2 | NP_038879.2 | 20 | 2864 | ||
GenBank mRNA | AF498362.1 | - | 20 | 2864 | ||
AK173034.1 | - | 19 | 2605 | |||
BC141401.1 | - | 20 | 2827 | 9056014 |