Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013852.2 | NP_038880.1 | 7 | 728 | ||
GenBank mRNA | AK030626.1 | - | 7 | 693 | ||
AK214630.1 | - | 4 | 359 | |||
BC032923.1 | - | 7 | 669 | 1430389 |