Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130458.1 | NP_001123930.1 | 10 | 1918 | ||
NM_001130459.1 | NP_001123931.1 | 10 | 1887 | |||
NM_015749.3 | NP_056564.1 | 10 | 1921 | |||
GenBank mRNA | AF090686.1 | - | 9 | 1848 | ||
AK133707.1 | - | 10 | 1883 | |||
AK143761.1 | - | 10 | 1889 | |||
AK147176.1 | - | 10 | 1892 | |||
AK161618.1 | - | 10 | 1888 | |||
AK170401.1 | - | 10 | 1907 | |||
AK202567.1 | - | 1 | 353 | |||
BC003720.1 | - | 9 | 1794 | 3601531 |