Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002294.2 | NP_001002294.1 | 4 | 511 | ||
NM_001319173.1 | NP_001306102.1 | 5 | 621 | |||
NM_001319174.1 | NP_001306103.1 | 3 | 322 | |||
NM_006894.5 | NP_008825.4 | 4 | 494 | |||
XM_011509345.2 | XP_011507647.1 | 5 | 642 | |||
GenBank mRNA | AK223166.1 | - | 4 | 503 | ||
AK296105.1 | - | 3 | 296 | |||
AK313197.1 | - | 4 | 484 | |||
BC032016.1 | - | 4 | 446 | 5175615 | ||
BM453951.1 | - | 4 | 523 | |||
KU178077.1 | - | 3 | 400 | |||
M83772.1 | - | 4 | 536 | |||
Z47552.1 | - | 4 | 493 |