Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025391.2 | NP_001020562.1 | 8 | 1191 | ||
NM_015752.3 | NP_056567.2 | 8 | 1194 | |||
GenBank mRNA | AF134893.1 | - | 8 | 1134 | ||
AJ131692.1 | - | 8 | 1192 | |||
AJ308625.1 | - | 8 | 870 | |||
AJ308626.1 | - | 8 | 1125 | |||
AK015885.1 | - | 7 | 952 | |||
AK016976.1 | - | 8 | 1172 | |||
AK047603.1 | - | 8 | 1064 | |||
AK156410.1 | - | 8 | 1156 | |||
BC048168.1 | - | 8 | 1164 | 6821477 | ||
BC056997.1 | - | 8 | 1164 | 6821477 |