Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015754.2 | NP_056569.2 | 5 | 461 | ||
GenBank mRNA | AF039563.1 | - | 5 | 380 | ||
AK017606.1 | - | 5 | 402 | |||
AK076944.1 | - | 5 | 456 | |||
BC011107.1 | - | 5 | 399 | 3982875 | ||
BC050201.1 | - | 5 | 397 | 30013927 | ||
BC147814.1 | - | 5 | 454 | 9087531 | ||
BC147815.1 | - | 5 | 454 | 9087532 |