Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016785.2 | NP_058065.2 | 8 | 749 | ||
GenBank mRNA | AF037043.1 | - | 8 | 658 | ||
AF037044.1 | - | 7 | 609 | |||
AF037045.1 | - | 7 | 609 | |||
AF046887.1 | - | 8 | 735 | |||
AK002335.1 | - | 8 | 656 | |||
BC021598.1 | - | 7 | 636 | 5098857 |