Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016852.2 | NP_058548.1 | 2 | 134 | ||
GenBank mRNA | AK004824.1 | - | 2 | 136 | ||
AK043037.1 | - | 2 | 108 | |||
AK079300.1 | - | 2 | 124 | |||
AK146572.1 | - | 2 | 136 | |||
AK150018.1 | - | 2 | 109 | |||
AK151368.1 | - | 2 | 126 | |||
AK162810.1 | - | 2 | 109 | |||
AK170317.1 | - | 2 | 135 | |||
AK170474.1 | - | 2 | 137 | |||
BC031908.1 | - | 2 | 97 | 5365118 | ||
BC055058.1 | - | 2 | 154 | 6529458 | ||
U40826.1 | - | 2 | 134 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK044015.1 | - | 124 | Chr. - - [-] on Build |