Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018871.3 | NP_061359.2 | 1, 2 | 327 | ||
GenBank mRNA | AB073970.1 | - | 1, 2 | 274 | ||
AF058799.1 | - | 1, 2 | 277 | |||
AK088847.1 | - | 1, 2 | 240 | |||
AK148618.1 | - | 1, 2 | 286 | |||
AK153307.1 | - | 1, 2 | 286 | |||
AK164356.1 | - | 1, 2 | 278 | |||
AK183704.1 | - | 1, 2 | 63 | |||
BC008129.1 | - | 1, 2 | 210 | 3498633 | ||
BY244024.1 | - | 1, 2 | 342 | |||
CT010208.1 | - | 1, 2 | 95 |