Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019668.3 | NP_062642.1 | 5 | 410 | ||
GenBank mRNA | AF089812.1 | - | 5 | 418 | ||
AF383148.1 | - | 5 | 418 | |||
AK133723.1 | - | 5 | 419 | |||
AK150417.1 | - | 5 | 393 | |||
AK153327.1 | - | 5 | 393 | |||
AK160527.1 | - | 5 | 405 | |||
AK160538.1 | - | 5 | 419 | |||
AK160545.1 | - | 5 | 417 | |||
AK163966.1 | - | 5 | 436 | |||
AK164074.1 | - | 5 | 419 | |||
BC026053.1 | - | 5 | 408 | 4012075 |