Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083921.1 | NP_001077390.1 | 3 | 587 | ||
NM_019705.3 | NP_062679.2 | 4 | 428 | |||
GenBank mRNA | AF124663.1 | - | 2 | 197 | ||
AK085559.1 | - | 3 | 576 | |||
AK088591.1 | - | 3 | 549 | |||
AK146176.1 | - | 3 | 546 | |||
AK166075.1 | - | 3 | 555 | |||
AK178495.1 | - | 2 | 155 | |||
AK181405.1 | - | 2 | 155 | |||
AK182273.1 | - | 2 | 155 | |||
AK210389.1 | - | 2 | 155 | |||
AK213806.1 | - | 2 | 155 | |||
BC034555.1 | - | 4 | 424 | 4192310 | ||
CX238505.1 | - | 3 | 587 |