Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019710.2 | NP_062684.2 | 1, 2 | 209 | ||
GenBank mRNA | AF047600.1 | - | 1, 2 | 209 | ||
AK013648.1 | - | 1, 2 | 164 | |||
AK017948.1 | - | 1, 2 | 159 | |||
AK088183.1 | - | 1, 2 | 146 | |||
BC003279.1 | - | 1, 2 | 146 | |||
BC025590.1 | - | 1, 2 | 152 | |||
BC036973.1 | - | 1, 2 | 162 | |||
BC131667.1 | - | 1, 2 | 162 | 40110822 |