Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003129.1 | 1 | 548 |
Gene Symbol: RNF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006913.3 | NP_008844.1 | 6 | 621 | ||
GenBank mRNA | AB056869.1 | - | 6 | 494 | ||
AK311859.1 | - | 6 | 504 | |||
AL555609.3 | - | 6 | 647 | |||
BC004155.2 | - | 1 | 498 | 2822537 | ||
BC020290.1 | - | 5 | 355 | |||
BC111392.1 | - | 6 | 615 | 6458319 | ||
BC119741.1 | - | 6 | 514 | 40119199 | ||
BC119742.1 | - | 6 | 514 | 40119200 | ||
BC127651.1 | - | 6 | 521 | 40119201 | ||
BC127652.1 | - | 6 | 521 | 40119202 | ||
BC148255.1 | - | 6 | 521 | 40119203 | ||
BT007105.1 | - | 6 | 494 | |||
BU599297.1 | - | 6 | 614 |