Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020275.4 | NP_064671.2 | 1 | 283 | ||
GenBank mRNA | AB031081.1 | - | 1 | 223 | ||
AF176833.1 | - | 1 | 95 | |||
AK050753.1 | - | 1 | 284 | |||
AK080069.1 | - | 1 | 274 | |||
AK145730.1 | - | 1 | 271 | |||
AK154993.1 | - | 1 | 263 | |||
BC065141.1 | - | 1 | 226 | 30608306 |