Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159502.1 | NP_001152974.1 | 2 | 366 | ||
NM_020494.3 | NP_065240.2 | 2 | 200 | |||
GenBank mRNA | AF214732.1 | - | 2 | 180 | ||
AK011012.1 | - | 2 | 144 | |||
AK036538.1 | - | 2 | 164 | |||
AK134339.1 | - | 2 | 178 | |||
AK149810.1 | - | 2 | 151 | |||
AK150778.1 | - | 2 | 152 | |||
AK152551.1 | - | 2 | 152 | |||
AK152928.1 | - | 2 | 152 | |||
AK166360.1 | - | 2 | 158 | |||
AK172557.1 | - | 2 | 151 | |||
BC043721.1 | - | 2 | 168 | |||
BC055048.1 | - | 2 | 130 | 6484820 | ||
BC055317.1 | - | 2 | 366 | 5696084 | ||
CJ141277.1 | - | 2 | 200 |