Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020601.2 | NP_065626.1 | 9 | 1040 | ||
GenBank mRNA | AK030547.1 | - | 9 | 1255 | ||
AK031937.1 | - | 5 | 501 | |||
AK036996.1 | - | 8 | 1484 | |||
AK038674.1 | - | 9 | 1041 | |||
BC043105.1 | - | 9 | 1163 | 6406579 |